missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium(V) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Vacuum deposition

Supplier:  Thermo Scientific Chemicals 04442422

 View more versions of this product

Catalog No. AA4442422



Description

Description

Vanadium(V) oxide is used in Vacuum deposition, catalysts for conversion of toluene to benzonitrile or propylene to acrylonitrile, as a detector material in bolometers and microbolometer arrays for thermal imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1750°C (decomposition)
690°C
Pieces
(metals basis)
MFCD00011457
14,9921
Soluble in water(8g/L).
O=[V](=O)O[V](=O)=O
181.88
CHEBI:30045
99.9%
3.357 g/mL
Vanadium (V) oxide
1314-62-1
100 g
O5V2
UN2862
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
GNTDGMZSJNCJKK-UHFFFAOYSA-N
dioxovanadiooxy(dioxo)vanadium
14814
181.88
Odorless

Chemical Identifiers

1314-62-1
181.88
GNTDGMZSJNCJKK-UHFFFAOYSA-N
14814
dioxovanadiooxy(dioxo)vanadium
O5V2
MFCD00011457
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride
CHEBI:30045
O=[V](=O)O[V](=O)=O
Safety and Handling

Safety and Handling

GHS H Statement
H372-H341-H361-H302-H332-H335
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P501c

H302+H332-H318-H335-H341-H351-H361d-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM PENTOXIDE

EINECSNumber : 215-239-8

RTECSNumber : YW2125000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only