missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium(V) oxide, 99.6% min, Alfa Aesar™

Manufacturer:  Alfa Aesar 1109409

 View more versions of this product

Catalog No. AA1109409



Specifications

Specifications

Vanadium(V) oxide
99.6% min
MFCD00011457
GNTDGMZSJNCJKK-UHFFFAOYSA-N
dioxovanadiooxy(dioxo)vanadium
14814
181.88
3.357g/mL
1314-62-1
O5V2
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride, vanadium oxide v2o5
O=[V](=O)O[V](=O)=O
181.878
CHEBI:30045
-10 Mesh
10g

Chemical Identifiers

1314-62-1
181.878
GNTDGMZSJNCJKK-UHFFFAOYSA-N
14814
dioxovanadiooxy(dioxo)vanadium
O5V2
MFCD00011457
vanadium v oxide, vanadium pentoxide, divanadium pentaoxide, divanadium pentoxide, vanadic anhydride, vanadin v oxide, vanadium oxide v2o5, vanadium pentaoxide, vanadic acid anhydride, vanadium oxide v2o5
CHEBI:30045
O=[V](=O)O[V](=O)=O
Safety and Handling

Safety and Handling

EINECSNumber : 215-239-8

Documents