Please login to your online account to display your discounted pricing

Vanadium Turnings, 2.5cm (1.0 in.) & down, 99.7% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Vanadium
CAS 7440-62-2
Physical Form Turnings, 2.5cm (1.0 in.) and down
Assay Percent Range 99.7% (Metals basis)
MDL Number MFCD00011453
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1042009

 
alfa aesar
1042009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 10g
Color Silver-gray
Density 6.11g/cm3
Physical Form Turnings, 2.5cm (1.0 in.) and down
Chemical Name or Material Vanadium
Assay Percent Range 99.7% (Metals basis)
MDL Number MFCD00011453
CAS 7440-62-2
Odor Odorless
Appearance Silver Gray turnings