missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 1106830

 View more versions of this product

Catalog No. AA1106830



Specifications

Specifications

Vanadium slug
250g
7440-62-2
V
00j9j9xkde
[V]
Odorless
23990
Slug
≈2.5g/slug
6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length
(metals basis)
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
vanadium
50.941
CHEBI:35170
99.8%

Chemical Identifiers

7440-62-2
50.941
LEONUFNNVUYDNQ-UHFFFAOYSA-N
23990
vanadium
V
MFCD00011453
00j9j9xkde
CHEBI:35170
[V]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents