Please login to your online account to display your discounted pricing

Vanadium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SV15500

 View more versions of this product

Catalog No. SV15-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 0°C
Purity Grade Certified Grade
Boiling Point 100°C
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Light Green
Density 1.0g/cm³
Vapor Pressure 14mmHg at 20°C
CAS 7647-01-0
Name Note 1000ppm ±1%
Chemical Name or Material Vanadium, Reference Standard Solution
MDL Number MFCD00011453
ChemAlert Storage Symbol White
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED VANADIUM STANDARD
CAS 7732-18-5
Concentration 990 to 1010ppm
CAS 1314-62-1

Specifications

Chemical Name or Material Vanadium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7647-01-0
CAS 1314-62-1
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00011453
Accessories
 

SDS & Safety Documents