Please login to your online account to display your discounted pricing

Vanadium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


Quantity 100mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLSV15100

 View more versions of this product

Catalog No. SV15-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Melting Point 0°C
Boiling Point 100°C
Quantity 100mL
DOT Information DOT Class 8, : Corrosive
Color Light Green
Physical Form Liquid
Vapor Pressure 14mmHg at 20°C
Density 1.0g/cm³
Name Note 1000ppm ±1%
ChemAlert Storage Symbol White
CAS 1314-62-1
CAS 7732-18-5
Chemical Name or Material Vanadium, Reference Standard Solution
Concentration 990 to 1010ppm
MDL Number MFCD00011453
CAS 7647-01-0
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED VANADIUM STANDARD

Specifications

Chemical Name or Material Vanadium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7647-01-0
CAS 1314-62-1
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00011453
Accessories
 

SDS & Safety Documents