Please login to your online account to display your discounted pricing

Vanadium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

$91.94 - $235.41

Chemical Identifiers

Chemical Name or Material Vanadium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7647-01-0
CAS 1314-62-1
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00011453
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SV15-500

  SDS
fisher scientific company
SV15500
Poly Bottle 500mL Each for $235.41
Case of 6 Each for $1,412.46

SV15-100

  SDS
fisher scientific company
FLSV15100
Poly Bottle 100mL Each for $91.94
Case of 6 Each for $551.64
Description & Specifications

Specifications

Melting Point 0°C
Purity Grade Certified Grade
Boiling Point 100°C
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Light Green
Density 1.0g/cm³
Vapor Pressure 14mmHg at 20°C
CAS 7647-01-0
Name Note 1000ppm ±1%
Chemical Name or Material Vanadium, Reference Standard Solution
MDL Number MFCD00011453
ChemAlert Storage Symbol White
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED VANADIUM STANDARD
CAS 7732-18-5
Concentration 990 to 1010ppm
CAS 1314-62-1
Safety and Handling