missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium powder, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1223418

 View more versions of this product

Catalog No. AA1223418



Specifications

Specifications

Vanadium
7440-62-2
99.5% (Metals basis)
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
vanadium
50.941
CHEBI:35170
Silver to Gray
50g
Silver Gray powder
V
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
[V]
Odorless
23990
Powder

Chemical Identifiers

7440-62-2
50.941
LEONUFNNVUYDNQ-UHFFFAOYSA-N
23990
vanadium
V
MFCD00011453
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
CHEBI:35170
[V]
Safety and Handling

Safety and Handling

H228-H315-H319-H335: Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-171-1

Documents