Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium plate, 3.2mm (0.13 in.) thick, annealed, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Vanadium plate
CAS 7440-62-2
Name Note 3.2mm (0.13 in.) thick, Annealed
MDL Number MFCD00011453
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11099FI

  SDS
alfa aesar™
11099FI
50x50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Vanadium plate
TSCA TSCA
Name Note 3.2mm (0.13 in.) thick, Annealed
Physical Form Plate
Assay 99.5%
Assay Percent Notes (metals basis)
MDL Number MFCD00011453
CAS 7440-62-2
Odor Odorless
Weight ≈48.9g/50x50mm
Quantity 50x50mm