Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium(IV,V) oxide, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Vanadium(IV,V) oxide
CAS 12037-42-2
Formula Weight 513.64
MDL Number MFCD00064848
UN Number UN3285
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2295609

 
alfa aesar™
2295609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2295618

 
alfa aesar™
2295618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 513.64
Chemical Name or Material Vanadium(IV,V) oxide
Odor Odorless
Assay 99.5%
Assay Percent Notes (metals basis)
MDL Number MFCD00064848
CAS 12037-42-2
UN Number UN3285
Color Blue-black
Melting Point ≈1500°C
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 3.91g/mL
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.