missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium(IV,V) oxide, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 2295618

 View more versions of this product

Catalog No. AA2295618



Specifications

Specifications

Vanadium(IV,V) oxide
99.5%
O13V6
UN3285
O=[V]=O.O=[V]=O.O=[V]=O.O=[V]=O.O=[V](=O)O[V](=O)=O
513.636
513.64
3.91g/mL
Black-Blue
50g
12037-42-2
(metals basis)
MFCD00064848
FTGZPVKRMCXHDZ-UHFFFAOYSA-N
dioxovanadiooxy(dioxo)vanadium;dioxovanadium
72720435
Crystalline Powder
Odorless
~1500°C

Chemical Identifiers

12037-42-2
513.636
FTGZPVKRMCXHDZ-UHFFFAOYSA-N
dioxovanadiooxy(dioxo)vanadium;dioxovanadium
O13V6
MFCD00064848
72720435
O=[V]=O.O=[V]=O.O=[V]=O.O=[V]=O.O=[V](=O)O[V](=O)=O
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.

SDS Documents