missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium(IV) Sulfate Oxide Hydrate, 99.9% (Metal Basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1109009

 View more versions of this product

Catalog No. AA1109009



Specifications

Specifications

Vanadium(IV) Sulfate Oxide Hydrate
99.9%, metal basis
MFCD00149933
DKCWBFMZNUOFEM-UHFFFAOYSA-L
oxovanadium(2+);sulfate;hydrate
16211496
Lumps
123334-20-3
H2O6SV
Vanadium oxysulfate hydrate
O.[O-]S(=O)(=O)[O-].O=[V+2]
181.011
163
10g

Chemical Identifiers

123334-20-3
181.011
DKCWBFMZNUOFEM-UHFFFAOYSA-L
16211496
O.[O-]S(=O)(=O)[O-].O=[V+2]
H2O6SV
MFCD00149933
Vanadium oxysulfate hydrate
oxovanadium(2+);sulfate;hydrate
Safety and Handling

Safety and Handling

Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste.

Harmful if swallowed.

DOTInformation : DOT Class: 6.1

EINECSNumber : 248-652-7

Documents