Learn More
Vanadium(IV) oxide phthalocyanine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03958702
Description
It is used as an organic light-emitting diode intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Vanadium(IV) oxide phthalocyanine | |
13930-88-6 | |
0.5 g | |
MFCD00041966 | |
oxyvanadium phthalocyanine, vanadium iv oxide phthalocyanine, vanadyl phthalocyanine, vopc, vanadyl iv phthalocyanine, vanadyl phthalocyanine, dye content >90 % | |
YRZZLAGRKZIJJI-UHFFFAOYSA-N | |
579.476 | |
579.45 |
Blue to Purple | |
Crystalline | |
C32H16N8OV | |
UN3285 | |
Insoluble in water. | |
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O=[V+2] | |
2735160 |
Chemical Identifiers
13930-88-6 | |
579.476 | |
YRZZLAGRKZIJJI-UHFFFAOYSA-N | |
2735160 |
C32H16N8OV | |
MFCD00041966 | |
oxyvanadium phthalocyanine, vanadium iv oxide phthalocyanine, vanadyl phthalocyanine, vopc, vanadyl iv phthalocyanine, vanadyl phthalocyanine, dye content >90 % | |
C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.O=[V+2] |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
EINECSNumber : 237-700-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only