Learn More
Vanadium(IV) oxide bis(2,4-pentanedionate), Thermo Scientific Chemicals
As a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment
$44.68 - $328.90
Chemical Identifiers
CAS | 3153-26-2 |
---|---|
Molecular Formula | C10H14O5V |
Molecular Weight (g/mol) | 265.16 |
MDL Number | MFCD00000032 |
InChI Key | JFHJZWAQYMGNBE-SUKNRPLKSA-L |
Synonym | vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide |
PubChem CID | 131674261 |
IUPAC Name | (Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate |
SMILES | O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA8111909
|
Thermo Scientific Chemicals
08111909 |
10 g |
Each for $44.68
|
|
|||||
AA8111918
|
Thermo Scientific Chemicals
08111918 |
50 g |
Each for $129.22
|
|
|||||
AA8111930
|
Thermo Scientific Chemicals
08111930 |
250 g |
Each for $328.90
|
|
|||||
Description
Employed as a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment. It is used in organic chemistry as a reagent in the epoxidation of allylic alcohols in combination with tert-butyl hydroperoxide (TBHP).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3153-26-2 | |
265.16 | |
JFHJZWAQYMGNBE-SUKNRPLKSA-L | |
131674261 | |
O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
C10H14O5V | |
MFCD00000032 | |
vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide | |
(Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate |
Specifications
140°C (13mmHg) | |
3153-26-2 | |
10 g | |
MFCD00000032 | |
vanadyl acetylacetonate, bis 2,4-pentanedionato vanadium iv oxide | |
JFHJZWAQYMGNBE-SUKNRPLKSA-L | |
(Z)-4-oxopent-2-en-2-olate;vanadium(2+);hydrate | |
131674261 | |
1.4 g/mL |
256°C to 259°C | |
Powder | |
C10H14O5V | |
UN3285 | |
Moderately soluble in acetone, ether and chloroform; Soluble in ethanol and benzene | |
O=[V++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
265.16 | |
265.16 | |
Vanadium (IV) oxide bis (2,4-pentanedionate) |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
EINECSNumber : 221-590-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only