Learn More
Vanadium(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02295718
Description
Vanadium(IV) oxide is used in chemical sensors, energy-conserving coatings, transparent conductors and switching materials. It acts as a stationary optical shutter, optical modulators, cameras and data storage. As infrared modulators, it is used for the missile guidance systems. Thin films made up of vanadium dioxide find applications in electro-optical switches, micro-optical switch, passive smart radiators and sunshields for spacecraft.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Vanadium(IV) oxide | |
12036-21-4 | |
50 g | |
O2V | |
vanadium dioxide, vanadium iv oxide, vanadium oxide vo2, vo2, vanadium iv oxide trace metals basis 10g | |
GRUMUEUJTSXQOI-UHFFFAOYSA-N | |
dioxovanadium | |
82849 | |
82.94 |
1967°C | |
Powder | |
99% (metals basis) | |
MFCD00049702 | |
Insoluble in water. | |
O=[V]=O | |
82.94 | |
CHEBI:30047 | |
4.339 |
Chemical Identifiers
12036-21-4 | |
82.94 | |
GRUMUEUJTSXQOI-UHFFFAOYSA-N | |
82849 | |
dioxovanadium |
O2V | |
MFCD00049702 | |
vanadium dioxide, vanadium iv oxide, vanadium oxide vo2, vo2, vanadium iv oxide trace metals basis 10g | |
CHEBI:30047 | |
O=[V]=O |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 234-841-1
TSCA : Yes
RUO – Research Use Only