Learn More
Vanadium(IV) fluoride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01154314
Description
Vanadium(IV) fluoride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Vanadium(IV) fluoride | |
Yellow to Brown | |
Powder | |
VF4 | |
UN2923 | |
Soluble in water (blue color), acetone (deep green color), and glacial acetic acid (blue-green color) | |
95% | |
2.975 g/mL |
325°C (decomposition) | |
10049-16-8 | |
25 g | |
MFCD00042554 | |
vanadium iv fluoride, acmc-20aja2, vanadium 4+ tetrafluoride, vanadium 4+ ion tetrafluoride, fluoride, fluoride, fluoride, vanadium salt, fluoride | |
126.94 | |
Moisture sensitive |
Chemical Identifiers
10049-16-8 | |
MFCD00042554 | |
vanadium iv fluoride, acmc-20aja2, vanadium 4+ tetrafluoride, vanadium 4+ ion tetrafluoride, fluoride, fluoride, fluoride, vanadium salt, fluoride |
Safety and Handling
GHS H Statement
H301-H331-H314-H318
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311+H331-H314
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 233-171-7
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only