Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium(IV) etioporphyrin III oxide

$587.64 - $587.64

Chemical Identifiers

Chemical Name or Material Vanadium(IV) etioporphyrin III oxide
CAS 15709-03-2
Formula Weight 543.61
MDL Number MFCD00058901
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39586MF

  SDS
alfa aesar™
39586MF
50mg Each for $587.64
Description & Specifications

Specifications

Formula Weight 543.61
Chemical Name or Material Vanadium(IV) etioporphyrin III oxide
Odor Odorless
Physical Form Crystalline
MDL Number MFCD00058901
CAS 15709-03-2
Color Purple
Quantity 50mg