Please login to your online account to display your discounted pricing

Vanadium(IV) chloride, 99+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  299690250

 View more versions of this product

Catalog No. AC299690250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0% (VCl4)
Physical Form Liquid
Formula Weight 192.75
Color Opaque reddish-brown
Quantity 25g
EINECS Number 231-561-1
TSCA TSCA
Density 1.8200g/mL
RTECS Number YW2625000
Chemical Name or Material Vanadium(IV) chloride
Iron (Fe) 200ppm max.
Name Note 99+%
Packaging Glass bottle
Assay Percent Range 99+%
Molecular Formula Cl4V
Linear Formula VCl4
MDL Number MFCD00061343
CAS 7632-51-1
Vanadium (V) 26.37% min. V(4+) (typically), 0.11% max. V(5+) (typically)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Vanadium(IV) chloride
Name Note 99+%
CAS 7632-51-1
Assay ≥99.0% (VCl4)
Assay Percent Range 99+%
Molecular Formula Cl4V
Linear Formula VCl4
Formula Weight 192.75
MDL Number MFCD00061343
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Contact with water liberates toxic gas.

SDS & Safety Documents