Learn More
Vanadium(III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals
$514.78 - $1975.53
Chemical Identifiers
CAS | 1314-34-7 |
---|---|
Molecular Formula | O3V2 |
Molecular Weight (g/mol) | 149.88 |
MDL Number | MFCD00011455 |
InChI Key | KFAFTZQGYMGWLU-UHFFFAOYSA-N |
Synonym | vanadium iii oxide, divanadium trioxide, vanadium oxide v2o3, vanadium 3 oxide, oxo oxovanadio oxy vanadium, vanadium trioxide, nonfused form, vanadium trioxide, v2o3, vanadium iii oxide trace metals basis |
PubChem CID | 518710 |
IUPAC Name | oxo(oxovanadiooxy)vanadium |
SMILES | O=[V]O[V]=O |
Description
Vanadium(III) oxide catalyzes the direct fluorination reactions. It is used in the coating of carbon nanotubes and as an electrode in lithium ion batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1314-34-7 | |
149.88 | |
KFAFTZQGYMGWLU-UHFFFAOYSA-N | |
518710 | |
O=[V]O[V]=O |
O3V2 | |
MFCD00011455 | |
vanadium iii oxide, divanadium trioxide, vanadium oxide v2o3, vanadium 3 oxide, oxo oxovanadio oxy vanadium, vanadium trioxide, nonfused form, vanadium trioxide, v2o3, vanadium iii oxide trace metals basis | |
oxo(oxovanadiooxy)vanadium |
Specifications
1970°C | |
Powder | |
99.7% (metals basis) | |
MFCD00011455 | |
Slightly soluble in water. | |
O=[V]O[V]=O | |
149.88 | |
149.88 | |
Vanadium(III) oxide |
1314-34-7 | |
100 g | |
O3V2 | |
vanadium iii oxide, divanadium trioxide, vanadium oxide v2o3, vanadium 3 oxide, oxo oxovanadio oxy vanadium, vanadium trioxide, nonfused form, vanadium trioxide, v2o3, vanadium iii oxide trace metals basis | |
KFAFTZQGYMGWLU-UHFFFAOYSA-N | |
oxo(oxovanadiooxy)vanadium | |
518710 | |
4.87 |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P321-P330-P332+P313-P361-P363-P501c
H302-H311-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 215-230-9
RTECSNumber : YW3050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only