missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium(III) oxide, 95%, Alfa Aesar™

Manufacturer:  Alfa Aesar 12301A1

 View more versions of this product

Catalog No. AA12301A1



Specifications

Specifications

1314-34-7
O3V2
vanadium iii oxide, vanadium iii oxide, divanadium trioxide, vanadium oxide v2o3, vanadium 3 oxide, oxo oxovanadio oxy vanadium, vanadium iii oxide, vanadium trioxide, nonfused form, vanadium trioxide, v2o3, vanadium iii oxide trace metals basis
O=[V]O[V]=O
149.88
149.88
Odorless
1kg
95%
MFCD00011455
KFAFTZQGYMGWLU-UHFFFAOYSA-N
oxo(oxovanadiooxy)vanadium
518710
Powder
Black

Chemical Identifiers

1314-34-7
149.88
KFAFTZQGYMGWLU-UHFFFAOYSA-N
518710
O=[V]O[V]=O
O3V2
MFCD00011455
vanadium iii oxide, vanadium iii oxide, divanadium trioxide, vanadium oxide v2o3, vanadium 3 oxide, oxo oxovanadio oxy vanadium, vanadium iii oxide, vanadium trioxide, nonfused form, vanadium trioxide, v2o3, vanadium iii oxide trace metals basis
oxo(oxovanadiooxy)vanadium
Documents