missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium(III) chloride, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1403918

 View more versions of this product

Catalog No. AA1403918



Specifications

Specifications

Vanadium(III) chloride
99% (metals basis)
MFCD00011454
Cl[V](Cl)Cl
157.292
157.3
50g
7718-98-1
Cl3V
HQYCOEXWFMFWLR-UHFFFAOYSA-K
trichlorovanadium
62647
-4 Mesh

Chemical Identifiers

7718-98-1
157.292
HQYCOEXWFMFWLR-UHFFFAOYSA-K
trichlorovanadium
Cl3V
MFCD00011454
62647
Cl[V](Cl)Cl
Safety and Handling

Safety and Handling

EINECSNumber : 231-744-6

Documents