Learn More
Vanadium(III) chloride, 97%, Thermo Scientific Chemicals
Vanadium(III) chloride, 97%, Cl3V, CAS Number-7718-98-1, Vanadium trichloride, 100g, 96% min. (Argentometry), Black to Blue, 3.0000g/mL, 231-744-6, 13,289, 157.3, GHS Signal Word: Danger
Supplier: Thermo Scientific Chemicals 197000050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Vanadium(III) chloride | |
99% | |
Black to Blue | |
5 g | |
Cl3V | |
MFCD00011454 | |
Vanadium trichloride | |
HQYCOEXWFMFWLR-UHFFFAOYSA-K | |
trichlorovanadium | |
62647 | |
97% | |
3 |
250.0°C | |
7718-98-1 | |
Powder | |
96% min. (Argentometry) | |
VCl3 | |
13,289 | |
Solubility in water: vigorous reaction. Other solubilities: soluble in abs.alcohol and ether | |
[Cl-].[Cl-].[Cl-].[V+3] | |
157.29 | |
157.3 | |
Glass bottle | |
3.0000g/mL |
Chemical Identifiers
7718-98-1 | |
157.29 | |
HQYCOEXWFMFWLR-UHFFFAOYSA-K | |
62647 | |
[Cl-].[Cl-].[Cl-].[V+3] |
Cl3V | |
MFCD00011454 | |
Vanadium trichloride | |
trichlorovanadium |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 231-744-6
RTECSNumber : YW2800000
TSCA : TSCA
RUO – Research Use Only