missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium(III) bromide, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8113504

 View more versions of this product

Catalog No. AA8113504



Specifications

Specifications

Vanadium(III) bromide
99.5% (metals basis)
MFCD00049695
ZOYIPGHJSALYPY-UHFFFAOYSA-K
tribromovanadium
83509
Crystalline
2g
13470-26-3
Br3V
vanadium bromide, vanadium tribromide, vanadium bromide vbr3, vanadium iii bromide, wln: va e3, vanadium iii bromide, vanadium iii bromide-v
[V](Br)(Br)Br
290.654
290.67
4.00^1^8 g/mL

Chemical Identifiers

13470-26-3
290.654
ZOYIPGHJSALYPY-UHFFFAOYSA-K
83509
[V](Br)(Br)Br
Br3V
MFCD00049695
vanadium bromide, vanadium tribromide, vanadium bromide vbr3, vanadium iii bromide, wln: va e3, vanadium iii bromide, vanadium iii bromide-v
tribromovanadium
Safety and Handling

Safety and Handling

EINECSNumber : 236-736-6

SDS Documents