missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium(III) 2,4-pentanedionate, typically 97%

Manufacturer:  Alfa Aesar™ 2294006

 View more versions of this product

Catalog No. AA2294006



Specifications

Specifications

Vanadium(III) 2,4-pentanedionate
5g
C5H8O2V
UN3285
CZFPVTPZFRXJDT-LNKPDPKZSA-N
(Z)-4-hydroxypent-3-en-2-one;vanadium
122198782
Powder
Air Sensitive
13476-99-8
MFCD00000033
Vanadium(III) acetylacetonate
CC(=CC(=O)C)O.[V]
151.059
348.27
97% (typically)

Chemical Identifiers

13476-99-8
151.059
CZFPVTPZFRXJDT-LNKPDPKZSA-N
122198782
CC(=CC(=O)C)O.[V]
C5H8O2V
MFCD00000033
Vanadium(III) acetylacetonate
(Z)-4-hydroxypent-3-en-2-one;vanadium
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.

TSCA : TSCA

SDS Documents