Learn More
Vanadium(III) 2,4-pentanedionate, typically 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02294014
Description
Vanadium(III) 2,4-pentanedionate is used as homogeneous catalyst, primary and secondary intermediates, Laboratory chemicals, and manufacture of substances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Vanadium(III) 2,4-pentanedionate | |
C15H21O6V | |
25 g | |
Air Sensitive | |
Soluble in water 2g/L,toluene,ethanol,acetone. | |
[V+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
348.27 | |
348.27 | |
Powder |
13476-99-8 | |
MFCD00000033 | |
UN3285 | |
Vanadium(III) acetylacetonate | |
ZVFSRLXFKIWGRT-UHFFFAOYSA-N | |
(Z)-4-hydroxypent-3-en-2-one;vanadium | |
122198782 | |
97% (typically) |
Chemical Identifiers
13476-99-8 | |
348.27 | |
ZVFSRLXFKIWGRT-UHFFFAOYSA-N | |
122198782 | |
[V+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
C15H21O6V | |
MFCD00000033 | |
Vanadium(III) acetylacetonate | |
(Z)-4-hydroxypent-3-en-2-one;vanadium |
Safety and Handling
GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
H301+H311-H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
EINECSNumber : 236-759-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only