missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium ICP Standard, 1mL = 10mg V (10,000ppm V), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPV10KN100

 View more versions of this product

Catalog No. PV10KN-100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Vanadium ICP Standard
77.0,20.24,0.77
V
[V]
50.941
CHEBI:35170
Poly natural
1 mL = 10 mg V
100mL
7440-62-2,7697-37-2,7732-18-5
80.0,21.07,0.8
LEONUFNNVUYDNQ-UHFFFAOYSA-N
vanadium
23990
Liquid
10,000ppm V
Yellow

Chemical Identifiers

7440-62-2
50.941
23990
vanadium
V
LEONUFNNVUYDNQ-UHFFFAOYSA-N
CHEBI:35170
[V]
Documents