Please login to your online account to display your discounted pricing

Vanadium ICP Standard, 1mL = 10mg V (10,000ppm V), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Vanadium ICP Standard
CAS 7732-18-5
CAS Min % 77.0
CAS Max % 80.0
CAS 7697-37-2
CAS Min % 20.24
CAS Max % 21.07
CAS 7440-62-2
CAS Min % 0.77
CAS Max % 0.8
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PV10KN-100

 
ricca chemical company
RIPV10KN100
Poly natural 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PV10KN-50

 
ricca chemical company
RIPV10KN50
Poly natural 50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PV10KN-500

 
ricca chemical company
RIPV10KN500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear yellow
Quantity 100mL
Chemical Name or Material Vanadium ICP Standard
CAS 7732-18-5
Concentration or Composition Notes 10,000ppm V
Equivalence 1 mL = 10 mg V
CAS 7697-37-2
CAS Min % 0.77
CAS Max % 21.07
Packaging Poly natural
CAS Min % 20.24
CAS Min % 77.0
CAS Max % 0.8
CAS 7440-62-2
CAS Max % 80.0