Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium gallide, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Vanadium gallide
CAS 12024-15-6
Formula Weight 222.55g/mol
MDL Number MFCD02091753
UN Number UN3285
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1751906

 
alfa aesar™
17519-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Mesh Size −100 Mesh
Quantity 5g
Melting Point 1720°C
Quantity 5g
Chemical Name or Material Vanadium gallide
CAS 12024-15-6
Physical Form Powder
Assay Percent Notes (metals basis)
Formula Weight 222.55g/mol
MDL Number MFCD02091753
UN Number UN3285
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.