missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium foil, 99.8% (metals basis), Alfa Aesar™

Form: approx. 3.87g/50x100mm

Manufacturer:  Alfa Aesar 13783GK

 View more versions of this product

Catalog No. AA13783GK



Specifications

Specifications

Vanadium
7440-62-2
99.8% (Metals basis)
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
vanadium
50.941
CHEBI:35170
Silver to Gray
100 x 200mm
Silver Gray foil
V
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
[V]
Odorless
23990
Foil, 0.127mm (0.005 in.) thick

Chemical Identifiers

7440-62-2
50.941
LEONUFNNVUYDNQ-UHFFFAOYSA-N
23990
vanadium
V
MFCD00011453
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
CHEBI:35170
[V]
Safety and Handling

Safety and Handling

EINECSNumber : 231-171-1

Documents