missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium foil, 99.5% (metal basis), Alfa Aesar™

Form: approx. 13g/50x150mm

Manufacturer:  Alfa Aesar 14633FZ

 View more versions of this product

Catalog No. AA14633FZ



Specifications

Specifications

Vanadium
7440-62-2
99.5% (Metals basis)
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
[V]
50.941
CHEBI:35170
Silver to Gray
50 x 150mm
Silver Gray foil
V
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
Odorless
vanadium
23990
Foil, 0.25mm (0.01 in.) thick

Chemical Identifiers

7440-62-2
50.941
LEONUFNNVUYDNQ-UHFFFAOYSA-N
23990
vanadium
V
MFCD00011453
dust, vanadium, elemental, ion, iii, 1+ , ion, vanadium, ion 3+, fume or dust, vanadium, ion v5+, unii-00j9j9xkde, 00j9j9xkde
CHEBI:35170
[V]
Safety and Handling

Safety and Handling

EINECSNumber : 231-171-1

SDS Documents