missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium foil, 1.0mm (0.04 in.) thick, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 11097GA

 View more versions of this product

Catalog No. AA11097GA



Specifications

Specifications

Vanadium foil
50 x 200mm
7440-62-2
V
00j9j9xkde
[V]
Odorless
23990
Foil
≈3.8g/25x25mm
1.0mm (0.04 in.) thick
(metals basis)
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
vanadium
50.941
CHEBI:35170
99.5%

Chemical Identifiers

7440-62-2
50.941
LEONUFNNVUYDNQ-UHFFFAOYSA-N
23990
vanadium
V
MFCD00011453
00j9j9xkde
CHEBI:35170
[V]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents