missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Vanadium foil, 0.075mm (0.003 in.) thick, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 44106FY

 View more versions of this product

Catalog No. AA44106FY



Specifications

Specifications

Vanadium foil
50 x 100mm
7440-62-2
V
00j9j9xkde
[V]
Odorless
23990
Foil
≈2.29g/50x100mm
0.075mm (0.003 in.) thick
(metals basis)
MFCD00011453
LEONUFNNVUYDNQ-UHFFFAOYSA-N
vanadium
50.941
CHEBI:35170
99.8%

Chemical Identifiers

7440-62-2
50.941
LEONUFNNVUYDNQ-UHFFFAOYSA-N
23990
vanadium
V
MFCD00011453
00j9j9xkde
CHEBI:35170
[V]
Safety and Handling

Safety and Handling

EINECSNumber : 231-171-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents