missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Vanadium carbide, 99% (metals basis), Alfa Aesar™

Chemical Identifiers

CAS 12070-10-9
Physical Form Powder
Assay Percent Range 99% metal basis
Molecular Formula VC (VC0?88 by X-Ray)
Formula Weight 62.95
MDL Number MFCD00049698
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1214114 SDS Alfa Aesar
1214114
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1214122 SDS Alfa Aesar
1214122
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

12070-10-9
99% metal basis
62.95
Powder
VC (VC0?88 by X-Ray)
MFCD00049698

Specifications

12070-10-9
99% metal basis
62.95
Odorless
25g
Powder
VC (VC0?88 by X-Ray)
MFCD00049698
Black