Please login to your online account to display your discounted pricing

Vanadate-Molybdate Reagent, for Phosphorus, LabChem™

$61.17 - $61.17

Chemical Identifiers

Chemical Name or Material Vanadate-Molybdate Reagent
CAS 7732-18-5
CAS Max % 83.71
CAS 7647-01-0
CAS Max % 13.66
CAS 12054-85-2
CAS Max % 2.5
CAS 7803-55-6
CAS Max % 0.13
Name Note For Phosphorus
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC266002

 
lab chem
LC266002
1L Each for $61.17
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 1235.86
Chemical Name or Material Vanadate-Molybdate Reagent
CAS Max % 2.5
Decomposition Information Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
CAS 7803-55-6
Color clear yellow-green solution
Quantity 1L
Sensitivity Passes Test
CAS Max % 0.13
Name Note For Phosphorus
Physical Form Liquid
Linear Formula (NH4)6Mo7O24·4H2O
CAS 7732-18-5
UN Number UN1789
CAS Max % 83.71
Identification Passes Test
CAS 7647-01-0
Packaging Poly Bottle
CAS Max % 13.66
Solubility Soluble in water
CAS 12054-85-2
Storage Information Room Temperature