Van de Graaff Accessories
Accessories for Electrostatic Demonstrations.
Manufacturer: Fisher Science Education N140
Includes: Metal hook
Description
- Tapered metal post at top of acrylic rod designed to fit into tapered female part on bottom of accessory
- With metal hook
Specifications
Van de Graff Electrostatic Generator | |
Mounting of accessory used in demonstration w/Van de Graff Electrostatic Generator | |
Metal hook | |
Insulating Stand |