Van de Graaff Accessories
Accessories for Electrostatic Demonstrations.
Manufacturer: Fisher Science Education N124
Description
Electric Plume
- Can be mounted to the dome of the Van de Graaff with its rubber vacuum cup
- Strips repel each other and stand up when charged
- Consists of a 9.2 in. dia. (23cm) metal wheel with radial spokes and discharge points
- Supported on a cone bearing
- Wheel spins rapidly as electrons escape from the points when held near an electrostatic generator
Specifications
Van de Graaff generator | |
Used as an accessory for electrostatic demonstrations | |
9.2 in. dia. Metal wheel with radial spokes and discharge points, stand | |
Electric whirl |