Please login to your online account to display your discounted pricing

Thermo Scientific™ Value Lab Refrigerators


Safely store samples at temperatures from 1° to 12°C with Thermo Scientific™ Value™ Laboratory Refrigerators.

Manufacturer: thermo scientific™  02LREETSA

 View more versions of this product

 Alert

These units are intended for basic laboratory use only. Storage of high-value, life science materials or clinical biologicals may require alarm, monitoring, back-up and temperature performance features. The need to add these features should be determined by your industrial hygenist or safety officer. These units are not suitable for corrosive environments.

Catalog No. 02LREETSA

  • / Each
 

Description & Specifications

Specifications

Doors 1 (1 compartment)
Certifications/Compliance cCSAus
Color White
For Use With (Application) Indoor General Storage Of Non-Critical Samples And Reagents
Typical Lifetime 10 Years
Min. Clearance 4 in. On Sides, 4 in. At Back, 1.25 in. On Top
Lighting Interior - No
Temperature (Setpoint) Adjustable Set Point - Yes
Defrost Automatic
Defrost Manual Defrost
Dimensions (L x W x H) Exterior 19 x 18.63 x 17.25 in. (48.3 x 47.3 x 43.8 cm)
Temperature Range (Metric) 1°C to 8°C
Temperature Range (Metric) 1° to 12°C
Voltage 115V
Voltage 120V
Electrical Requirements 115 V/60 Hz
Control Mechanical
Warranty 13 Months Parts and Labor
Current 1.2 Amps / 83 Watts
Capacity 1.8 cu. ft. / 51L
Capacity 1.8 Cubic Feet/51 Liters
Casters No Leveling Feet
Casters None
Compressor Bottom Mounted
Door Count 1 (1 Compartment)
Door Count 1(1 compartment)
Door Style Hinged, Opens Left to Right, Reversible
Exterior Finish Enameled Steel
Dimensions (D x W x H) Interior 14 x 16 x 15.5 in. (35.6 x 40.6 x 39.4cm)
Hertz 60Hz
Interior Finish ABS Plastic
Insulation Polyurethane Foam
Model 02LREETSA
Net Weight (Metric) 20kg
Plug Type NEMA 5-15P
Plug Type Yes/Three prong NEMA 5-15P, 1.828 Meters (6 Feet)
Shipping Weight (English) 55 lb.
Shipping Weight (English) 75 lb.
Shelves 1 Fixed Interior Shelf, Storage On Bottom
Shelves 1 Removable Wire Shelf, Storage On Bottom Of Unit
Refrigerant R134a, 100% CFC Free
Refrigerant R134A,CFCFree
Shipping Weight (Metric) 24.9kg
Shipping Weight (Metric) 34kg
Type General Purpose Refrigerator
Amperage 1.2A
Capacity (English) 1.8 cu. ft.
Capacity (Metric) 51L
Net Weight (English) 47 lb.
Access Security Door lock
Depth (English) Interior 12 in. Upper; 9 in. Lower
Height (English) Interior 15.5 in.
Height (Metric) Exterior 49.5cm
Width (English) Exterior 20 in.
Width (English) Interior 16.6 in.
Width (Metric) Interior 42.2cm
Capacity (Metric) 51L
Depth (Metric) Interior 30.5cm Upper; 22.9cm Lower
Capacity 1.8 cu. ft. / 51L
Height (English) Exterior 19.5 in.
Length (English) Exterior 20.5 in.
Capacity (English) 1.8 cu. ft.
Width (Metric) Exterior 50.8cm
Chamber Material Enameled steel, White

Safely store samples at temperatures from 1° to 12°C with Thermo Scientific™ Value™ Laboratory Refrigerators, featuring adjustable temperature control and manual defrost.

  • Adjustable shelves or baskets included
  • Environmentally friendly, CFC-free insulation and coolant
  • Factory tested prior to shipment
  • NRTL tested
  • Easy-to-clean, durable finish 

Ordering Information:

Not available outside North America.