Learn More
Thermo Scientific™ Value Lab Refrigerators
Safely store samples at temperatures from 1° to 12°C with Thermo Scientific™ Value™ Laboratory Refrigerators.
Manufacturer: Thermo Scientific™ 05LREETSA
Alert:
These units are intended for basic laboratory use only. Storage of high-value, life science materials or clinical biologicals may require alarm, monitoring, back-up and temperature performance features. The need to add these features should be determined by your industrial hygenist or safety officer. These units are not suitable for corrosive environments.
Description
Safely store samples at temperatures from 1° to 12°C with Thermo Scientific™ Value™ Laboratory Refrigerators, featuring adjustable temperature control and manual defrost.
- Adjustable shelves or baskets included
- Environmentally friendly, CFC-free insulation and coolant
- Factory tested prior to shipment
- NRTL tested
- Easy-to-clean, durable finish
Ordering Information:
Not available outside North America.
Specifications
Door lock | |
White | |
1.0 Amps/160 Watts | |
155.7L | |
23.5 x 23.63 x 33.5 in. (60 x 60 x 85.1 cm) | |
NEMA 5-15P | |
General Purpose Refrigerator | |
1(1 compartment) | |
ABS Plastic | |
54.5kg | |
13 Months Parts and Labor | |
10 Years | |
Interior - Yes | |
1.33A | |
Bottom Mounted | |
Polyurethane Foam | |
R134a, 100% CFC Free | |
100 lb. | |
Manual |
cULus | |
Mechanical Dial Thermostat/ Inside, Upper Right Side | |
5.5 cu. ft. | |
Enameled Steel | |
17.5 x 20 x 27.75 in. (44.5 x 50.8 x 70.5cm) | |
1° to 12°C | |
115V | |
115 V/60 Hz | |
100 lb. | |
160w | |
Indoor General Storage Of Non-Critical Samples And Reagents | |
2 in. On Sides, 2 in. At Back, 2 in. On Top | |
Adjustable | |
2 Adjustable Feet In Front, 2 Fixed Pads In Rear | |
60Hz | |
05LREETSA | |
3 Adjustable Shelves, Storage On Bottom Of Unit | |
45.3kg | |
Hinged, Opens Left to Right, Reversible |