Please login to your online account to display your discounted pricing

Valinomycin, Tocris

Selective K+ ionophore

$76.90 - $293.30

Chemical Identifiers

Chemical Name or Material Valinomycin
Assay Percent Range >98%
Molecular Formula C54H90N6O18
Formula Weight 1111.32
Synonym Cyclo(D-α-hydroxyisovaleryl-D-valyl-L-lactoyl-L-valyl-D-α-hydroxyisovaleryl-D-valyl-L-lactoyl-L-valyl-D-α-hydroxyisovaleryl-D-valyl-L-lactoyl-L-valyl)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

337350

 
r & d systems
RDS337350
50mg Each for $293.30

337310

 
r & d systems
RDS337310
10mg Each for $76.90
Description & Specifications

Specifications

Formula Weight 1111.32
Quantity 50mg
Chemical Name or Material Valinomycin
Assay Percent Range >98%
Molecular Formula C54H90N6O18
CAS 2001-95-8
Synonym Cyclo(D-α-hydroxyisovaleryl-D-valyl-L-lactoyl-L-valyl-D-α-hydroxyisovaleryl-D-valyl-L-lactoyl-L-valyl-D-α-hydroxyisovaleryl-D-valyl-L-lactoyl-L-valyl)
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C