missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Valinomycin, 10mg/mL in DMSO, sterile-filtered, Alfa Aesar™

$82.91 - $352.95

Chemical Identifiers

CAS 2001-95-8
Molecular Formula C54H90N6O18
Molecular Weight (g/mol) 1111.338
MDL Number MFCD00005114
InChI Key FCFNRCROJUBPLU-CVZFCLRKSA-N
PubChem CID 131668508
IUPAC Name (3S,6S,12R,15S,18S,21R,24R,27S,30S,33R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nona(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
SMILES CC1C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)O1)C(C)C)C(C)C)C(C)C)C)C(C)C)C(C)C)C(C)C)C)C(C)C)C(C)C)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67410XF
View Documents
Thermo Fisher Scientific
J67410XF
1mL Each for $82.91
 
AAJ674108EQ
View Documents
Thermo Fisher Scientific
J674108EQ
5 x 1mL Pack for $352.95
 
Specifications

Chemical Identifiers

2001-95-8
1111.338
FCFNRCROJUBPLU-CVZFCLRKSA-N
(3S,6S,12R,15S,18S,21R,24R,27S,30S,33R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nona(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
C54H90N6O18
MFCD00005114
131668508
CC1C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)O1)C(C)C)C(C)C)C(C)C)C)C(C)C)C(C)C)C(C)C)C)C(C)C)C(C)C)C(C)C

Specifications

2001-95-8
MFCD00005114
Plastic bottle
CC1C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)O1)C(C)C)C(C)C)C(C)C)C)C(C)C)C(C)C)C(C)C)C)C(C)C)C(C)C)C(C)C
1111.338
Liquid
Valinomycin
C54H90N6O18
78657
FCFNRCROJUBPLU-CVZFCLRKSA-N
(3S,6S,12R,15S,18S,21R,24R,27S,30S,33R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nona(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
131668508
1mL
Safety and Handling

Safety and Handling

P210-P264b-P270-P280-P301+P310-P302+P352-P312-P330-P361-P363-P370+P378q-P501c

H227-H301+H311

EINECSNumber : 217-896-6

RTECSNumber : YV9468000

TSCA : Yes

Recommended Storage : Store at -20°C