Please login to your online account to display your discounted pricing

Alfa Aesar™ Valinomycin, 10mg/mL in DMSO, sterile-filtered

Chemical Identifiers

Chemical Name or Material Valinomycin
CAS 2001-95-8
Name Note 10mg/mL in DMSO, sterile-filtered
MDL Number MFCD00005114
Beilstein 78657
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ674108EQ

  SDS
alfa aesar™
J674108EQ
5 x 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Valinomycin
Health Hazard 2 GHS H Statement
H302-H312-H227
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Physical Form Liquid
Name Note 10mg/mL in DMSO, sterile-filtered
CAS 2001-95-8
Health Hazard 1 Warning
MDL Number MFCD00005114
Health Hazard 3 GHS P Statement
P210-P280-P301+P312-P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Quantity 5 x 1mL
Storage Note 1 -30° to -10°C
Beilstein 78657
RTECS Number YV9468000
Safety and Handling

Warning

GHS P Statement
P210-P280-P301+P312-P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H227
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.

-30° to -10°C