missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Valethamate bromide, 99%

$70.93 - $134.16

Chemical Identifiers

CAS 90-22-2
Molecular Formula C19H32BrNO2
Molecular Weight (g/mol) 386.374
MDL Number MFCD00031617
InChI Key CEJGGHKJHDHLAZ-UHFFFAOYSA-M
Synonym valethamate bromide, ediposin, epidosin, epidozin, resitan, murel, valethamate bromide jan, n,n-diethyl-n-methyl-2-3-methyl-2-phenylpentanoyl oxy ethanaminium bromide, diethyl methyl 2-3-methyl-2-phenylvaleryloxy ethyl ammonium bromide, 2-diethylaminoethyl 2-phenyl-3-methylvalerate methyl bromide
PubChem CID 7010
IUPAC Name diethyl-methyl-[2-(3-methyl-2-phenylpentanoyl)oxyethyl]azanium;bromide
SMILES CCC(C)C(C1=CC=CC=C1)C(=O)OCC[N+](C)(CC)CC.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6262206 View Documents Alfa Aesar™
J6262206
5g Each for $70.93
 
AAJ6262214 View Documents Alfa Aesar™
J6262214
25g Each for $134.16
 
Specifications

Chemical Identifiers

90-22-2
386.374
CEJGGHKJHDHLAZ-UHFFFAOYSA-M
7010
CCC(C)C(C1=CC=CC=C1)C(=O)OCC[N+](C)(CC)CC.[Br-]
C19H32BrNO2
MFCD00031617
valethamate bromide, ediposin, epidosin, epidozin, resitan, murel, valethamate bromide jan, n,n-diethyl-n-methyl-2-3-methyl-2-phenylpentanoyl oxy ethanaminium bromide, diethyl methyl 2-3-methyl-2-phenylvaleryloxy ethyl ammonium bromide, 2-diethylaminoethyl 2-phenyl-3-methylvalerate methyl bromide
diethyl-methyl-[2-(3-methyl-2-phenylpentanoyl)oxyethyl]azanium;bromide

Specifications

White
5g
C19H32BrNO2
14,9906
CEJGGHKJHDHLAZ-UHFFFAOYSA-M
diethyl-methyl-[2-(3-methyl-2-phenylpentanoyl)oxyethyl]azanium;bromide
7010
Powder
Valethamate bromide
120°C to 127°C
90-22-2
MFCD00031617
valethamate bromide, ediposin, epidosin, epidozin, resitan, murel, valethamate bromide jan, n,n-diethyl-n-methyl-2-3-methyl-2-phenylpentanoyl oxy ethanaminium bromide, diethyl methyl 2-3-methyl-2-phenylvaleryloxy ethyl ammonium bromide, 2-diethylaminoethyl 2-phenyl-3-methylvalerate methyl bromide
CCC(C)C(C1=CC=CC=C1)C(=O)OCC[N+](C)(CC)CC.[Br-]
386.374
386.37
99%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : BP7600000