missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Valerophenone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1052536
Description
Valerophenone is used as intermediates of liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsValerophenone is used as intermediates of liquid crystals.
Solubility
Insoluble in water.
Notes
 Incompatible with strong oxidizing agents, acids, bases, plastics. Store at room temperature with proper ventilation.
Specifications
Valerophenone | |
-9°C | |
244°C to 245°C | |
99% | |
1.5143 | |
500 g | |
valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci | |
XKGLSKVNOSHTAD-UHFFFAOYSA-N | |
1-phenylpentan-1-one | |
66093 | |
162.23 |
1009-14-9 | |
0.978 | |
102°C (215°F) | |
C11H14O | |
MFCD00009480 | |
1907717 | |
Insoluble in water. | |
CCCCC(=O)C1=CC=CC=C1 | |
162.23 | |
CHEBI:36812 | |
99% |
Chemical Identifiers
1009-14-9 | |
162.23 | |
XKGLSKVNOSHTAD-UHFFFAOYSA-N | |
66093 | |
1-phenylpentan-1-one |
C11H14O | |
MFCD00009480 | |
valerophenone, butyl phenyl ketone, pentanophenone, 1-phenyl-1-pentanone, 1-pentanone, 1-phenyl, unii-f27q043nt1, n-butyl phenyl ketone, 1-benzoylbutane, valerophenone, 8ci | |
CHEBI:36812 | |
CCCCC(=O)C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : 213-767-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only