Please login to your online account to display your discounted pricing

Valeric anhydride, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  422450050

 View more versions of this product

Catalog No. AC422450050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 186.25
Physical Form Liquid
Quantity 5g
Color Clear colorless to yellow
Density 0.9200g/mL
Beilstein 02, IV, 874
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula (CH3(CH2)3CO)2O
CAS 2082-59-9
Chemical Name or Material Valeric anhydride
Refractive Index 1.4200 to 1.4220 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C10H18O3
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
MDL Number MFCD00009477
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Valeric anhydride
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H18O3
Linear Formula (CH3(CH2)3CO)2O
Formula Weight 186.25
MDL Number MFCD00009477
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents