Learn More
Valeraldehyde, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 149562500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Valeraldehyde | |
110-62-3 | |
0.8070g/mL | |
12°C | |
Glass bottle | |
1.3900 to 1.3950 | |
MFCD00007026 | |
01, 676 | |
15, 10088 | |
Solubility in water: 14g/L (20°C). Other solubilities: soluble in alcohol, ether, propylene glycol and, many organic solvents | |
CCCCC=O | |
86.13 | |
0.6 mPa.s (20°C) | |
86.13 |
98% | |
-92.0°C | |
103.0°C | |
96% min. (GC) | |
C5H10O | |
CH3(CH2)3CHO | |
250 mL | |
0.807 | |
valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde | |
HGBOYTHUEUWSSQ-UHFFFAOYSA-N | |
pentanal | |
8063 | |
CHEBI:84069 | |
97% |
Chemical Identifiers
110-62-3 | |
86.13 | |
HGBOYTHUEUWSSQ-UHFFFAOYSA-N | |
8063 | |
pentanal |
C5H10O | |
MFCD00007026 | |
valeraldehyde, n-pentanal, n-valeraldehyde, valeric aldehyde, valeral, valeryl aldehyde, amylaldehyde, butyl formal, amyl aldehyde, valeric acid aldehyde | |
CHEBI:84069 | |
CCCCC=O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 203-784-4
RTECSNumber : YV3600000
TSCA : TSCA
RUO – Research Use Only