Please login to your online account to display your discounted pricing
Valence Shell Electron Pair Repulsion Models
Manufacturer: fisher science education  VSP-1
 View more versions of this product
Catalog No. S65370
  • / Each
Description & Specifications
Specifications
Includes White 1-peg Hydrogen atom centers, blue 4-peg Nitrogen atom centers, red 4-peg Oxygen atom centers, green 4-peg Halogen atom centers, black 4-peg Carbon atom centers, yellow 4-peg Sulfur atom centers, 1.25 in. PVC, clear bonds (single bonds), 1.625 in. PVC, clear bonds (double bonds), background information booklet, lab procedure, student response form and answer key
For Use With (Application) Illustrates molecular geometry, resonance and molecule polarity
Type Valence shell electron pair repulsion model