Please login to your online account to display your discounted pricing

Alfa Aesar™ Val-Glu-Pro-Ile-Pro-Tyr

For cellular and molecular biology applications

Specifications

Product Type Val-Glu-Pro-Ile-Pro-Tyr
Molecular Weight 716.82
Storage Requirements -30° to -10°C
Sequence Immunostimulating Peptide, human
Format Lyophilized powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66362LB0

 
alfa aesar™
J66362LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66362MCR

 
alfa aesar™
J66362MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Val-Glu-Pro-Ile-Pro-Tyr
Molecular Weight 716.82
Storage Requirements -30° to -10°C
Sequence Immunostimulating Peptide, human
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC