MilliporeSigma™ Vacuum Filtering Side-Arm Flask
Designed for use in vacuum filtration procedures with classic Glass Filter Holders
$90.00 - $369.00
Specifications
For Use With (Application) Vacuum Filtration Using Classic Filter Holders
Material Borosilicate glass
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Description Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Description Price Quantity  
XX1014705
View Documents
MilliporeSigma™
XX1014705
1 L Threaded side-arm
Each for $90.00
XX1014744
View Documents
MilliporeSigma™
XX1014744
4 L Threaded side-arm
Each for $369.00
Description
Description
Designed for use in vacuum filtration procedures with classic Glass Filter Holders
  • Side arm connects to vacuum source with 3/8 in. I.D. hose
  • 1 L and 4 L flasks accept No. 8 perforated stopper
  • 125 mL flask accepts No. 5 stopper
Specifications
Specifications
Vacuum Filtration Using Classic Filter Holders
Borosilicate glass
SDS