Please login to your online account to display your discounted pricing

Whatman™ Vacu-Guard Filter: PROMO OFFER

PTFE membrane retains 99.9% of airborne particles ≥0.1μm

$160.30 - $160.30

Specifications

Material (Membrane) PTFE
Filtration Area (Metric) 16cm2
For Use With (Application) Isolate Infectious Materials
View More Specs
Products
Catalog Number Mfr. No. Inlet Style Outlet Style Price Quantity    

0974475PM

 
whatman™
67225000PM
1/4-3/8 in. stepped hose barb 1/4-3/8 in. stepped hose barb Pack of 10 Each for $160.30

0974476PM

 
whatman™
67225001PM
3/8-1/2 in. stepped hose barb 3/8-1/2 in. stepped hose barb Pack of 10 Each for $160.30
Description & Specifications

Specifications

Material (Membrane) PTFE
Filtration Area (Metric) 16cm2
For Use With (Application) Isolate Infectious Materials

  • Installed inline
  • Prevent fluid and aerosol contamination of vacuum pumps or aspiration suction systems
  • Eliminate hazardous exhaust
  • Disposable
  • Filter area: for 50mm diameter