missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

V5 tag Mouse, Clone: TCM5, eBioscience™

Mouse Monoclonal Antibody

$117.48 - $315.48

Specifications

Antigen V5 tag
Clone TCM5
Host Species Murine
Applications Immunocytochemistry,Western Blot
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013554 Life Technologies
LS14679680
25μg Each for $117.48
5013555 Life Technologies
LS14679682
100μg Each for $315.48
Specifications

Description

This TCM5 monoclonal antibody recognizes recombinant proteins containing a V5 epitope tag. The V5 epitope is naturally present on the P and V proteins of the paramyxovirus, SV5. TCM5 specifically recognizes the sequence GKPIPNPLLGLDST.
Description & Specifications

Specifications

V5 tag
Murine
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TCM5
Immunocytochemistry,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL